missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100956
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR33.
Orphan receptor; could be a chemoattractant receptor.
Specifications
| GPR33 | |
| Polyclonal | |
| Unconjugated | |
| GPR33 | |
| G protein-coupled receptor 33 (gene/pseudogene); G protein-coupled receptor 33 pseudogene; GPR33; Probable G-protein coupled receptor 33 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2856, 299007 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q49SQ1 | |
| GPR33 | |
| A synthesized peptide derived from human GPR33(Accession Q49SQ1), corresponding to amino acid residues I148-R198. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction