missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533692
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%).
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR31 is a high-affinity receptor for 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (12-S-HETE). 12-(S)-HETE is an arachidonic acid metabolite secreted by platelets and tumor cells, and known to induce endothelial cells retraction allowing invasive cell access to the subendothelial matrix, which is a critical step for extravasation or metastasis. Ligand-binding leads to activation of MAPK and NF-kappa-B pathways.
Specifications
| GPR31 | |
| Polyclonal | |
| Unconjugated | |
| GPR31 | |
| 12-(S)-HETE acid receptor; 12-(S)-HETE receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE) receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid receptor; 12-HETER; bA517H2.2 (G protein-coupled receptor 31); G protein-coupled receptor 31; GPR31; G-protein coupled receptor 31; HETER; HETER1; hydroxyeicosatetraenoic (HETE) acid receptor 1; probable G-protein coupled receptor 31; TCP10C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O00270 | |
| GPR31 | |
| Synthetic 19 amino acid peptide from 2nd extracellular domain of human GPR31. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction