missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00196
This item is not returnable.
View return policy
Description
GPR3 Polyclonal specifically detects GPR3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| GPR3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ACCA, G protein-coupled receptor 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2827 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_005272.1 | |
| GPR3 | |
| Peptide with sequence C-LQRHLLPASH corresponding to internal region according to NP_005272.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is not expected to cross-react with GPR6 and GPR12. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction