missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567863
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
G protein coupled receptors (GPCRs) are a large family of receptors that have a similar structure characterized by 7 transmembrane domains. Activation of GPCRs by extracellular stimuli such as neurotransmitters, hormones, or light induces an intracellular signaling cascade mediated by heterotrimeric GTP-binding proteins, or G proteins. The 375 amino acid GPCR GPR27 protein contains several features of the GPCR family, including 7 transmembrane domains, potential N linked glycosylation sites in the N terminal extracellular domain, and conserved cysteine residues, which may form a disulfide bridge, in the first and second extracellular loops.
Specifications
| GPR27 | |
| Polyclonal | |
| Unconjugated | |
| GPR27 | |
| G protein-coupled receptor 27; GPR27; Probable G-protein coupled receptor 27; SREB1; Super conserved receptor expressed in brain 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14761, 2850, 65275 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54897, Q9JJH3, Q9NS67 | |
| GPR27 | |
| Synthesized peptide derived from human GPR27. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction