missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR22 Monoclonal Antibody (455108)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561447
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR22 is an orphan G-protein-coupled receptor, belonging to the class A rhodopsin-like GPCRs. GPR22 is highly expressed in several human brain regions, heart and coronary arteries. GPR22 expression is also observed in normal pancreas and in pancreatic, colorectal, prostate and liver cancer.
Specifications
| GPR22 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q99680 | |
| GPR22 | |
| HEK293 human embryonic kidney cell line transfected with human GPR22, Met1-Asp433. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 455108 | |
| Unconjugated | |
| GPR22 | |
| 2900068K05Rik; AW061316; G protein-coupled receptor 22; GPR22; G-protein coupled receptor 22; MGC129847; probable G-protein coupled receptor 22; tcag7.108 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction