missing translation for 'onlineSavingsMsg'
Learn More

GPR22 Antibody (455108) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30178123 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178123 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30178123 Supplier Novus Biologicals Supplier No. FAB4500JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR22 Monoclonal antibody specifically detects GPR22 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR22
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 455108
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human GPR22, Met1-Asp433, Accession # Q99680
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2845
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.