missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR22 Antibody (455108), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4500CL1
This item is not returnable.
View return policy
Description
GPR22 Monoclonal antibody specifically detects GPR22 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| GPR22 | |
| Monoclonal | |
| CoraFluor 1 | |
| G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22 | |
| HEK293 human embryonic kidney cell line transfected with human GPR22, Met1-Asp433, Accession # Q99680 | |
| 0.1 mL | |
| GPCR | |
| 2845 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 455108 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction