missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570784
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 93%.
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids b chain (active). HuFSH induces maturation of Graafian follicles of the ovary, promotes development of germinal cells in males and activates cytosolic tyrosine kinase. The HuFSH is purified by proprietary chromatographic techniques.
Specifications
| GPR21 | |
| Polyclonal | |
| Unconjugated | |
| GPR21 | |
| C230004C13Rik; G protein-coupled receptor 21; G protein-coupled receptor GPR21; Gpr21; Probable G-protein coupled receptor 21 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99679 | |
| GPR21 | |
| synthetic peptide directed towards the C-terminal region of human GPR21. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction