missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA120406
This item is not returnable.
View return policy
Description
PA1-20406 detects G Protein-Coupled Receptor GPR19 in human samples.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR19 is an orphan receptor found primarily in the central nervous system. GPR19 was also found in ovary and testis and in human embryonic stem cells. GPR19 has been postulated to play a role in cancer development and increased levels of GPR19 mRNA were found in metastatic melanoma, small cell lung carcinoma and pancreas islet cell carcinoma.
Specifications
| GPR19 | |
| Polyclonal | |
| Unconjugated | |
| GPR19 | |
| G protein-coupled receptor 19; GPR19; GPR-NGA; Probable G-protein coupled receptor 19 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100066597, 100350488, 2842, 540647 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15760 | |
| GPR19 | |
| Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human GPR19. | |
| 25 μg | |
| Primary | |
| Human, Rabbit, Monkey, Bovine, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction