missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GPR19 Antibody (6A1D8), Novus Biologicals™
Mouse Monoclonal Antibody
£226.00 - £389.00
Specifications
| Antigen | GPR19 |
|---|---|
| Clone | 6A1D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30028493
|
Novus Biologicals™
NBP3-27167-0.1ml |
0.1 mL |
£389.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029012
|
Novus Biologicals™
NBP3-27167-0.025ml |
0.025 mL |
£226.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR19 Monoclonal antibody specifically detects GPR19 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| GPR19 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| GPCR | |
| PBS | |
| 2842 | |
| IgG2a | |
| Protein G purified |
| 6A1D8 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| G protein-coupled receptor 19, GPR-NGA, probable G-protein coupled receptor 19 | |
| Purified recombinant fragment of human GPR19 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title