missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR183 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114732
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EBI2.
GPR183 was identified by the up-regulation of its expression upon Epstein-Barr virus infection of primary B lymphocytes. GPR183 is predicted to encode a G protein-coupled receptor that is most closely related to the thrombin receptor. Expression of this gene was detected in B-lymphocyte cell lines and lymphoid tissues but not in T-lymphocyte cell lines or peripheral blood T lymphocytes. The function of this gene is unknown.
Specifications
| GPR183 | |
| Polyclonal | |
| Unconjugated | |
| GPR183 | |
| EBI 2; Ebi2; EBV-induced gene 2; EBV-induced G-protein coupled receptor 2; EBV-induced G-protein coupled receptor 2 homolog; Epstein-Barr virus induced gene 2; Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor); epstein-Barr virus-induced G-protein coupled receptor 2; Epstein-Barr virus-induced G-protein coupled receptor 2 homolog; G protein-coupled receptor 183; GPR183; G-protein coupled receptor 183; RP11-461N23.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1880, 321019 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P32249, Q3U6B2 | |
| GPR183 | |
| A synthesized peptide derived from human GPR183(Accession P32249), corresponding to amino acid residues I212-V262. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction