missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR180 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106842
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR180.
This gene encodes a protein that is a member of the G protein-coupled receptor superfamily. This protein is produced predominantly in vascular smooth muscle cells and may play an important role in the regulation of vascular remodeling.
Specifications
| GPR180 | |
| Polyclonal | |
| Unconjugated | |
| GPR180 | |
| AB041545; E130016I23Rik; G protein-coupled receptor 180; GPR180; Integral membrane protein GPR180; intimal thickness-related receptor; ITR; MNCb-3029 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 160897, 58245 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86V85, Q8BPS4 | |
| GPR180 | |
| A synthesized peptide derived from human GPR180(Accession Q86V85), corresponding to amino acid residues I249-S299. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction