missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR177/WLS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59013
This item is not returnable.
View return policy
Description
GPR177/WLS Polyclonal specifically detects GPR177/WLS in Human samples. It is validated for Western Blot.
Specifications
| GPR177/WLS | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C1orf139, DKFZp686I0788, EVIchromosome 1 open reading frame 139, FLJ23091, G protein-coupled receptor 177, GPR177wls, Integral membrane protein GPR177, MGC131760, MGC14878, MRP, Protein evenness interrupted homolog, protein wntless homolog, Putative NF-kappa-B-activating protein 373, putative NFkB activating protein 373, wntless homolog (Drosophila) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q5T9L3 | |
| WLS | |
| Synthetic peptides corresponding to GPR177(G protein-coupled receptor 177) The peptide sequence was selected from the middle region of GPR177. Peptide sequence DIRLVGIHQNGGFTKVWFAMKTFLTPSIFIIMVWYWRRITMMSRPPVLLE. | |
| 100 μL | |
| Signal Transduction | |
| 79971 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction