missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR177 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567054
This item is not returnable.
View return policy
Description
Immunogen sequence: LAYRDDAFAEW TEMAHERVPR KLKCTFTSPK TPEHEGRYYE CDVLPFMEIG SVAHKFYLLN IRLPVNEKKK INVGIGEIKD Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.
Regulates Wnt proteins sorting and secretion in a feedback regulatory mechanism. This reciprocal interaction plays a key role in the regulation of expression, subcellular location, binding and organelle-specific association of Wnt proteins. Plays also an important role in establishment of the anterior-posterior body axis formation during development. [UniProt].
Specifications
| GPR177 | |
| Polyclonal | |
| Unconjugated | |
| WLS | |
| 5031439A09Rik; AI173978; AI987742; C1orf139; evenness interrupted homolog; EVI; G protein-coupled receptor 177; Gpr177; Integral membrane protein GPR177; mig-14; MRP; Protein evenness interrupted homolog; protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373; UNQ85/PRO18667; WLS; Wnt ligand secretion mediator; wntless homolog; wntless homolog (Drosophila); wntless Wnt ligand secretion mediator | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79971 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T9L3 | |
| WLS | |
| Recombinant Human GPR177. Recombinant protein control fragment (Product #RP-105477). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction