missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: DIRLVGIHQN GGFTKVWFAM KTFLTPSIFI IMVWYWRRIT MMSRPPVLLE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Regulates Wnt proteins sorting and secretion in a feedback regulatory mechanism. This reciprocal interaction plays a key role in the regulation of expression, subcellular location, binding and organelle-specific association of Wnt proteins. Plays also an important role in establishment of the anterior-posterior body axis formation during development. [UniProt].
Specifications
Specifications
| Antigen | GPR177 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | WLS |
| Gene Accession No. | Q5T9L3 |
| Gene Alias | 5031439A09Rik; AI173978; AI987742; C1orf139; evenness interrupted homolog; EVI; G protein-coupled receptor 177; Gpr177; Integral membrane protein GPR177; mig-14; MRP; Protein evenness interrupted homolog; protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373; UNQ85/PRO18667; WLS; Wnt ligand secretion mediator; wntless homolog; wntless homolog (Drosophila); wntless Wnt ligand secretion mediator |
| Gene Symbols | WLS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
