Learn More
Invitrogen™ GPR173 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102093
Product Code. 16678564
  • £449.00 / 100µL
Description
Antibody detects endogenous levels of total GPR173.
This gene encodes a member of the G-protein coupled receptor 1 family. This protein contains 7 transmembrane domains and conserved cysteine residues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR173
Polyclonal
Unconjugated
GPR173
3230401K02Rik; G protein-coupled receptor 173; G protein-coupled receptor SREB3; GPR173; G-protein coupled receptor 173; Probable G-protein coupled receptor 173; RP1-290F12.1; SREB3; Super conserved receptor expressed in brain 3
Rabbit
Affinity chromatography
RUO
54328, 64021, 70771
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6PI62, Q9JJH2, Q9NS66
GPR173
A synthesized peptide derived from human GPR173(Accession Q9NS66), corresponding to amino acid residues M254-V304.
100 μL
Primary
Human, Mouse, Rat