missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR173 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106835
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR173.
This gene encodes a member of the G-protein coupled receptor 1 family. This protein contains 7 transmembrane domains and conserved cysteine residues.
Specifications
| GPR173 | |
| Polyclonal | |
| Unconjugated | |
| GPR173 | |
| 3230401K02Rik; G protein-coupled receptor 173; G protein-coupled receptor SREB3; GPR173; G-protein coupled receptor 173; Probable G-protein coupled receptor 173; RP1-290F12.1; SREB3; Super conserved receptor expressed in brain 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54328, 64021, 70771 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6PI62, Q9JJH2, Q9NS66 | |
| GPR173 | |
| A synthesized peptide derived from human GPR173(Accession Q9NS66), corresponding to amino acid residues M254-V304. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction