missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GPR172A Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Packungsgröße:
0.10mL
Spezifikation
Spezifikation
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 79581 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 20 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | D15Ertd747e, G protein-coupled receptor 172 A, GPCR41FLJ11856, hRFT3, PAR1PERV-A receptor 1, Porcine endogenous retrovirus A receptor 1, Protein GPR172A, putative G-protein coupled receptor GPCR41, RFT3, riboflavin transporter 3 |
| Gene Symbol | SLC52A2 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49382. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur