missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR171 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533662
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Dog, Bovine, Panda (100%) Marmoset, Bat, Rabbit (94%) Elephant, Platypus (88%).
GPR171 is a protein coding gene. Orphan receptor.
Specifications
| GPR171 | |
| Polyclonal | |
| Unconjugated | |
| GPR171 | |
| A630006E05; F730001G15Rik; G protein-coupled receptor 171; Gpr171; G-protein coupled receptor H963; H963; platelet activating receptor homolog; probable G-protein coupled receptor 171 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29909, 485716, 688737, 767929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O14626, Q3ZBK9 | |
| GPR171 | |
| Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR171. | |
| 50 μg | |
| Primary | |
| Human, Rat, Canine, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction