missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Antibody (CL10474), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07993-25ul
This item is not returnable.
View return policy
Description
GPR17 Monoclonal antibody specifically detects GPR17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GPR17 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10474 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: KRLKGPPPSFEGKTNESSLS | |
| 25 μg | |
| GPCR, Neuroscience | |
| 2840 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction