Learn More
GPR17 Antibody (CL10474) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07993MFV500
Product Code. 30301699
  • £574.00 / 0.10mL
Description
GPR17 Monoclonal antibody specifically detects GPR17 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR17
Monoclonal
mFluor Violet 500 SE
DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor
This antibody was developed using a synthetic peptide derived from Q13304, with the exact immunogen sequence remaining proprietary.
0.1 mL
GPCR, Neuroscience
2840
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL10474
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified