missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR164 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-02972-100UL
This item is not returnable.
View return policy
Description
GPR164 Polyclonal specifically detects GPR164 in Human samples. It is validated for Western Blot.
Specifications
| GPR164 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| D-GPCR, Dresden-G-protein-coupled receptor, FLJ13581, GPR136, GPR164, G-protein coupled receptor 164, MGC24137, olfactory receptor 51E1, Olfactory receptor 52A3, olfactory receptor OR11-15, olfactory receptor, family 51, subfamily E, member 1, olfactory receptor, family 51, subfamily E, member 1 pseudogene, olfactory receptor, family 52, subfamily A, member 3 pseudogene, OR51E1P, OR52A3P, POGR, prostate overexpressed G protein-coupled receptor, Prostate-overexpressed G protein-coupled receptor, Prostate-specific G protein-coupled receptor 2, PSGR2 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human GPR164 (NP_689643.2). | |
| 100 μL | |
| GPCR | |
| 143503 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction