missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR162 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570902
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%; Zebrafish: 79%.
This gene was identified upon genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be mainly expressed in the brain; however, its function is not known. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| GPR162 | |
| Polyclonal | |
| Unconjugated | |
| GPR162 | |
| A-2; BC031437; G protein-coupled receptor 162; gene rich cluster, A; gene-rich cluster gene A protein; GPR162; GRCA; probable G-protein coupled receptor 162 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27239 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q16538 | |
| GPR162 | |
| synthetic peptide directed towards the N-terminal region of human GPR162. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction