missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR162 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533655
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Dog, Elephant, Panda, Horse, Pig (100%).
This gene was identified upon genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be mainly expressed in the brain; however, its function is not known. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| GPR162 | |
| Polyclonal | |
| Unconjugated | |
| GPR162 | |
| A-2; BC031437; G protein-coupled receptor 162; gene rich cluster, A; gene-rich cluster gene A protein; GPR162; GRCA; probable G-protein coupled receptor 162 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100146294, 100736664, 27239, 486719 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q16538 | |
| GPR162 | |
| Synthetic 17 amino acid peptide from 1st extracellular domain of human GPR162. | |
| 50 μg | |
| Primary | |
| Canine, Horse, Human, Monkey, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction