missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR158 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102092
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR158.
Orphan receptor.
Specifications
| GPR158 | |
| Polyclonal | |
| Unconjugated | |
| GPR158 | |
| 5330427M13Rik; G protein-coupled receptor 158; Gpr158; KIAA1136; mKIAA1136; Probable G-protein coupled receptor 158 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 241263, 57512 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5T848, Q8C419 | |
| GPR158 | |
| A synthesized peptide derived from human GPR158(Accession Q5T848), corresponding to amino acid residues L10-P60. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction