missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR158 Antibody (1027651), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB102861PECY7
This item is not returnable.
View return policy
Description
GPR158 Monoclonal antibody specifically detects GPR158 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR158 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1027651 | |
| Flow Cytometry | |
| G protein-coupled receptor 158, KIAA1136FLJ37801, probable G-protein coupled receptor 158, RP11-59G22.1 | |
| Human embryonic kidney cell HEK293-derived human GPR158 protein, Ala24-Gln411, Accession # Q5T848 | |
| 0.1 mL | |
| GPCR | |
| 57512 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction