missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR157 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555949
This item is not returnable.
View return policy
Description
Immunogen sequence: RLFSLCCCCC SSQPPTKSPA GTPKAPAPSK PGESQESQGT P Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 59%.
Orphan receptor that promotes neuronal differentiation of radial glial progenitors (RGPs). The activity of this receptor is mediated by a G(q)-protein that activates a phosphatidylinositol-calcium second messenger.
Specifications
| GPR157 | |
| Polyclonal | |
| Unconjugated | |
| GPR157 | |
| F730108M23Rik; FLJ12132; G protein-coupled receptor 157; GPR157; G-protein coupled receptor 157; novel 7 transmembrane receptor (Secretin family) domain containing protein; probable G-protein coupled receptor 157 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80045 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5UAW9 | |
| GPR157 | |
| Recombinant protein corresponding to Human GPR157. Recombinant protein control fragment (Product #RP-95165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction