missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR153 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120591
This item is not returnable.
View return policy
Description
Positive test controls include: Mouse lung. Immunogen sequence: MSDERRLPGS AVGWLVCGGL SLLANAWGIL SVGAKQKKWK PLEFLLCTLA ATHMLNVAVP IATYSVVQLR RQRPDFEWNE GLCKVFVSTF YTLTLATCFS.
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR153 (G protein-coupled receptor 153), also known as PGR1, is a 609 amino acid multi-pass membrane protein that functions as an orphan receptor and belongs to the GPR1 family. The gene encoding GPR153 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.
Specifications
| GPR153 | |
| Polyclonal | |
| Unconjugated | |
| GPR153 | |
| 1110065N12Rik; AI449320; G protein-coupled receptor 153; G protein-coupled receptor PGR1; GPR153; G-protein coupled receptor PGR1; Pgr1; probable G-protein coupled receptor 153 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100129, 387509, 619550 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6NV75, Q8K0Z9 | |
| GPR153 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GPR153 (NP_997253.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction