missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR152 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106834
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR152.
G protein-coupled receptors (GPRs or GPCRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, are members of the largest protein family and play a role in many different stimulus-response pathways. G protein-coupled receptors mediate extracellular signals into intracellular signals (G protein activation). They respond to a wide variety of signaling molecules, including hormones, neurotransmitters and other proteins and peptides. GPR proteins are usually integral seven pass membrane proteins with some conserved amino acid regions. GPR-152 (G proteincoupled receptor 152) is a 511 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family. Encoded by a gene on human chromosome 11q13.2 and mouse chromosome 19 A, GPR-152 may function as an orphan receptor.
Specifications
| GPR152 | |
| Polyclonal | |
| Unconjugated | |
| GPR152 | |
| A930009H15Rik; G protein-coupled receptor 152; G protein-coupled receptor PGR5; Gm673; Gpr152; G-protein coupled receptor PGR5; PGR5; Probable G-protein coupled receptor 152; putative G-protein coupled receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 269053, 390212 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BXS7, Q8TDT2 | |
| GPR152 | |
| A synthesized peptide derived from human GPR152(Accession Q8TDT2), corresponding to amino acid residues M1-P45. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction