missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR150 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102063
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR150.
G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 150 (GPR150), also designated PGR11 and seven transmembrane helix receptor, is a member of the rhodopsin family of GCPRs and is involved in signal transduction. GPR150 is expressed in various human tissues, including normal small intestine, skeletal muscle, kidney and tonsil, as well as cancerous blood, bladder, placenta and parathyroid.
Specifications
| GPR150 | |
| Polyclonal | |
| Unconjugated | |
| GPR150 | |
| C030001A19Rik; G protein-coupled receptor 150; Gpr150; G-protein coupled receptor PGR11; GPRv40; Pgr11; probable G-protein coupled receptor 150; RGD1564729; seven transmembrane helix receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 285601 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NGU9 | |
| GPR150 | |
| A synthesized peptide derived from human GPR150(Accession Q8NGU9), corresponding to amino acid residues L49-G99. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction