Learn More
Invitrogen™ GPR15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532802
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon (100%); Monkey (95%); Marmoset, Mouse, Bat, Bovine, Elephant, Panda, Rabbit (89%); Dog, Hamster, Horse, Pig (84%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
GPR15 expression has been reported in CD4(+) T lymphocytes, alveolar macrophages, and the basal surface of the small intestinal epithelium. ESTs have been isolated from normal brain and kidney cancer libraries.
Specifications
| GPR15 | |
| Polyclonal | |
| Unconjugated | |
| GPR15 | |
| 4933439K08Rik; BoB; brother of Bonzo; G protein-coupled receptor 15; GPR15; Gpr-15; G-protein coupled receptor 15; MGC126828; MGC126830 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P49685 | |
| GPR15 | |
| Synthetic 19 amino acid peptide from C-terminal cytoplasmic domain of human GPR15. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.