missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR15 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76466H
This item is not returnable.
View return policy
Description
GPR15 Polyclonal antibody specifically detects GPR15 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| GPR15 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| BOB, Brother of Bonzo, G protein-coupled receptor 15, G-protein coupled receptor 15, MGC126828, MGC126830 | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human GPR15. The immunogen is located within the first 50 amino acids of GPR15. Amino Acid Squence: YYATSPNSDIRETHSH | |
| 0.1 mL | |
| GPCR | |
| 2838 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction