missing translation for 'onlineSavingsMsg'
Learn More

GPR15 Antibody (367902), HRP, Novus Biologicals™

Product Code. 18619473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18619473 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18619473 Supplier Novus Biologicals Supplier No. FAB3654H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR15 Monoclonal antibody specifically detects GPR15 in Human samples. It is validated for Flow Cytometry, CyTOF-reported
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR15
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 367902
Conjugate HRP
Dilution Flow Cytometry, CyTOF-reported
Formulation PBS
Gene Alias BOB, Brother of Bonzo, G protein-coupled receptor 15, G-protein coupled receptor 15, MGC126828, MGC126830
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR15, Met1-Leu360, Accession # P49685
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2838
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.