missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR15 Antibody (367902), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3654Y
This item is not returnable.
View return policy
Description
GPR15 Monoclonal antibody specifically detects GPR15 in Human samples. It is validated for Flow Cytometry, CyTOF-reported
Specifications
| GPR15 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 367902 | |
| Flow Cytometry, CyTOF-reported | |
| BOB, Brother of Bonzo, G protein-coupled receptor 15, G-protein coupled receptor 15, MGC126828, MGC126830 | |
| NS0 mouse myeloma cell line transfected with human GPR15, Met1-Leu360, Accession # P49685 | |
| 0.1 mL | |
| GPCR | |
| 2838 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction