missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR146 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10154-100UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
GPR146 Polyclonal specifically detects GPR146 in Mouse samples. It is validated for Western Blot.
Tekniske data
| GPR146 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| G protein-coupled receptor 146, G protein-coupled receptor PGR8, PGR8G-protein coupled receptor PGR8, probable G-protein coupled receptor 146 | |
| The immunogen is a synthetic peptide directed towards the middle terminal region of mouse GPR146 (NP_001033792.1). Peptide sequence ERALPRTYMASVYNTRHVCGFVWGGAVLTSFSSLLFYICSHVSSRIAECA | |
| 100 μg | |
| GPCR | |
| 115330 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur