missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR146 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114737
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR146.
GPR146 is an Orphan-U GPCR with an unknown ligand, but is likely to be a chemokine receptor. ESTs have been isolated from B-cell/lung/testis, kidney, liver/spleen, lung, prostate, and spleen libraries.
Specifications
| GPR146 | |
| Polyclonal | |
| Unconjugated | |
| GPR146 | |
| BC003323; G protein-coupled receptor 146; Gpr146; G-protein coupled receptor PGR8; PGR8; Probable G-protein coupled receptor 146; RGD1560731 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 115330, 498153 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96CH1 | |
| GPR146 | |
| A synthesized peptide derived from human GPR146(Accession Q96CH1), corresponding to amino acid residues V192-Q242. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction