missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR143 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15719940 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15719940 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15719940 Supplier Invitrogen™ Supplier No. PA551984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CSLGFQSPRK EIQWESLTTS AAEGAHPSPL MPHENPASGK VSQVGGQTSD EALSMLSEGS DASTIEIHTA SESCNKNEGD PALPT Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.

Ocular albinism type 1 protein is a conserved integral membrane protein with seven transmembrane domains. It is expressed in the eye and epidermal melanocytes. Defects in GPR143 are the cause of ocular albinism type 1 (OA1); also known as Nettleship-Falls type ocular albinism. OA1 is an X-linked disorder characterized by severe impairment of visual acuity, retinal hypopigmentation and the presence of macromelanosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR143
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR143
Gene Accession No. P51810
Gene Alias G protein-coupled receptor 143; GPR143; G-protein coupled receptor 143; homolog of human ocular albinism 1 (Nettleship-Falls); MOA1; NYS6; OA1; ocular albinism 1; ocular albinism type 1 protein; Ocular albinism type 1 protein homolog; RGD1565799
Gene Symbols GPR143
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR143. Recombinant protein control fragment (Product #RP-92329).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4935
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.