Learn More
GPR141 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28096-1-AP-20UL
This item is not returnable.
View return policy
Description
The GPR141 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GPR141. This antibody recognizes human, mouse antigen. The GPR141 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| GPR141 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| G protein coupled receptor 141, GPR141, PGR13 | |
| 20 μL | |
| 353345 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |