missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR141 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595714
This item is not returnable.
View return policy
Description
Immunogen sequence: VTTMAVINLV VVHSVFLLTV PFRLTYLIKK TWMFGLPFCK FVSAMLHIHM YLTFLFYVVI LVTRYLIFFK CKDKVEFYRK LHAVAASAGM WTLVIVIVVP L; Positive Samples: Mouse lung, Rat lung; Cellular Location: Cell membrane, Multi-pass membrane protein.
GPR141 is a member of the rhodopsin family of G protein-coupled receptors (GPRs).
Specifications
| GPR141 | |
| Polyclonal | |
| Unconjugated | |
| Gpr141 | |
| G protein-coupled receptor 141; GPR141; G-protein coupled receptor PGR13; Pgr13; probable G-protein coupled receptor 141 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291179, 353346 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TQP0 | |
| Gpr141 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human GPR141 (NP_0013169221). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction