missing translation for 'onlineSavingsMsg'
Learn More

GPR139 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17268083 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17268083 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17268083 Supplier Thermo Scientific Supplier No. PA5115319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GPR139 Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR139
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GPR139
Gene Accession No. Q6DWJ6, Q80UC8
Gene Alias G protein-coupled receptor 139; G protein-coupled receptor PGR3; g(q)-coupled orphan receptor GPRg1; Gm495; Gpr139; GPRG1; G-protein coupled receptor 139; G-protein-coupled receptor PGR3; orphan G protein-coupled receptor TRHRL2; PGR3; Probable G-protein coupled receptor 139
Gene Symbols GPR139
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR139(Accession Q6DWJ6), corresponding to amino acid residues H187-F237.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124274, 209776
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.