missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR137C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102078
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR137C.
GPR137C is a protein coding gene.
Specifications
| GPR137C | |
| Polyclonal | |
| Unconjugated | |
| GPR137C | |
| 6330416L11Rik; G protein-coupled receptor 137C; G protein-coupled receptor TM7SF1L2; Gm908; GPR137C; Integral membrane protein GPR137C; TM7SF1L2; Transmembrane 7 superfamily member 1-like 2 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 283554, 70713 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N3F9 | |
| GPR137C | |
| A synthesized peptide derived from human GPR137C(Accession Q8N3F9), corresponding to amino acid residues L256-V306. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction