Learn More
GPR137B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16496-1-AP-20UL
 View more versions of this product
Product Code. 16876144
  • £135.00 / 20µL
Description
GPRs are characterized by the presence of 7 transmembrane domains. G Protein-Coupled Receptor (GPCR) TM7SF1 is a member of this family (subfamily: orphan U) and its expression has been reported in adult kidney and heart, but not in fetal kidney.
Specifications
Specifications
GPR137B
Polyclonal
Unconjugated
GPR137B
GPR137B, TM7SF1
Rabbit
Antigen Affinity Chromatography
RUO
7107
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60478
GPR137B
GPR137B Fusion Protein Ag9633
20 μL