missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR137B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533628
This item is not returnable.
View return policy
Description
GPR137B Polyclonal Antibody for ICC/IF, IHC (P)
GPRs are characterized by the presence of 7 transmembrane domains. G Protein-Coupled Receptor (GPCR) TM7SF1 is a member of this family (subfamily: orphan U) and its expression has been reported in adult kidney and heart, but not in fetal kidney.
Specifications
| GPR137B | |
| Polyclonal | |
| Unconjugated | |
| GPR137B | |
| G protein-coupled receptor 137B; GPR137B; Integral membrane protein GPR137B; RP5-985L19.1; TM7SF1; transmembrane 7 superfamily member 1 (upregulated in kidney); transmembrane 7 superfamily member 1 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100050643, 100154578, 100343157, 100764875, 289287, 421511, 479194, 613826, 7107, 83924 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O60478, Q8BNQ3 | |
| GPR137B | |
| Synthetic 16 amino acid peptide from 2nd cytoplasmic domain of human GPR137B / TM7SF1. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction