missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR133 Polyclonal Antibody

GPR133 Polyclonal Antibody
Brand: Thermo Scientific PA5106839
This item is not returnable.
View return policy
Description
GPR133 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| GPR133 | |
| Polyclonal | |
| Unconjugated | |
| ADGRD1 | |
| Adgrd1; adhesion G protein-coupled receptor D1; adhesion G-protein coupled receptor D1; DKFZp434B1272; E230012M21Rik; FLJ16770; G protein-coupled receptor 133; GPR133; G-protein coupled receptor 133; G-protein coupled receptor PGR25; MGC138512; MGC138514; Pgr25; probable G-protein coupled receptor 133 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 243277, 283383, 689257 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6QNK2, Q80T32 | |
| ADGRD1 | |
| A synthesized peptide derived from human ADGRD1(Accession Q6QNK2), corresponding to amino acid residues E466-S516. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction