missing translation for 'onlineSavingsMsg'
Learn More

GPR128 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£385.00 - £587.00

Specifications

Antigen GPR128
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18316715
View Documents
Novus Biologicals
NBP3-17498-25UL
25 μg
£385.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18316966
View Documents
Novus Biologicals
NBP3-17498-100UL
100 μg
£587.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GPR128 Polyclonal antibody specifically detects GPR128 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

GPR128
Immunofluorescence
Unconjugated
Rabbit
GPCR
PBS, pH 7.2, 40% glycerol
84873
IgG
Affinity purified
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Polyclonal
Purified
RUO
Human
FLJ14454, FLJ16382, FLJ29035, G protein-coupled receptor 128, MGC142011, MGC163260, probable G-protein coupled receptor 128
This antibody was developed against Recombinant Protein corresponding to amino acids: STSSSSTPTEFCRNGGTWENGRCICTEEWKGLRCTIANFCENSTYMGFTFARIPVGRYGPSLQTCGKDTPNAGNPMAVRLCSLSLYGEIELQKVTIGNCNENLETLEKQVKDVTAPLNNISSEVQILTSDANKLTAEN
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.