Learn More
Invitrogen™ GPR128 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139831
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: STSSSSTPTE FCRNGGTWEN GRCICTEEWK GLRCTIANFC ENSTYMGFTF ARIPVGRYGP SLQTCGKDTP NAGNPMAVRL CSLSLYGEIE LQKVTIGNCN ENLETLEKQV KDVTAPLNNI SSEVQILTSD ANKLTAEN Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 70%.
GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.
Specifications
| GPR128 | |
| Polyclonal | |
| Unconjugated | |
| ADGRG7 | |
| 9130020O16Rik; Adgrg7; adhesion G protein-coupled receptor G7; adhesion G-protein coupled receptor G7; FLJ14454; FLJ16382; FLJ29035; G protein-coupled receptor 128; GPR128; G-protein coupled receptor 128; MGC142011; MGC163260; probable G-protein coupled receptor 128 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96K78 | |
| ADGRG7 | |
| Recombinant protein corresponding to Human ADGRG7. Recombinant protein control fragment (Product #RP-108929). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.