missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR126 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115318
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR126.
GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
Specifications
| GPR126 | |
| Polyclonal | |
| Unconjugated | |
| ADGRG6 | |
| 1190004A11Rik; ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; adhesion G-protein coupled receptor G6; AI449247; APG1; AW045736; bge; big ears; developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; Gm222; gpr126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; probable G-protein coupled receptor 126; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57211 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q86SQ4 | |
| ADGRG6 | |
| A synthesized peptide derived from human ADGRG6(Accession Q86SQ4), corresponding to amino acid residues E435-N485. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction