missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR126 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572210
This item is not returnable.
View return policy
Description
GPR126 Polyclonal Antibody for Western Blot
GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
Specifications
| GPR126 | |
| Polyclonal | |
| Unconjugated | |
| ADGRG6 | |
| 1190004A11Rik; ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; adhesion G-protein coupled receptor G6; AI449247; APG1; AW045736; bge; big ears; developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; Gm222; gpr126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; probable G-protein coupled receptor 126; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 561970 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| C6KFA3 | |
| ADGRG6 | |
| KLH conjugated synthetic peptide between 1135-1169 amino acids from the C-terminal region of DANRE gpr126. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction