missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR126 Monoclonal Antibody (1044430)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560895

Product Code. 30534269

  • £324.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPR126
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q86SQ4
ADGRG6
Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437.
100 μg
Primary
Human
Antibody
IgG2a
Flow Cytometry
1044430
Unconjugated
ADGRG6
1190004A11Rik; ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; adhesion G-protein coupled receptor G6; AI449247; APG1; AW045736; bge; big ears; developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; Gm222; gpr126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; probable G-protein coupled receptor 126; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR
Mouse
Protein A/G
RUO
57211
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.