missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR123 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GPR123 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16879554
|
Proteintech
20075-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16869554
|
Proteintech
20075-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to GPR123.
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR123 (G protein-coupled receptor 123) is a 1,279 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Existing as two alternatively spliced isoforms, GPR123 functions as an orphan receptor that is expressed in adult and fetal brain, and in adult spinal cord. The gene encoding GPR123 maps to human chromosome 10q26.3 and mouse chromosome 7 F4.Specifications
| GPR123 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q86SQ6 | |
| 84435 | |
| Peptide | |
| Primary | |
| -20°C | |
| ADGRA1 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| G protein coupled receptor 123, GPR123, KIAA1828 | |
| ADGRA1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title